accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOFHOOH

CAS RN 2096336-07-9

IUPAC Name: {4-[(benzyloxy)carbonyl]-2-fluorophenyl}boronic acid
Synonyms: 4-[(benzyloxy)carbonyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 274.05
Molecular Formula: C14H12BFO4
InChi Key: DOERPIDBVIPDOC-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=C(C=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results