accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHNH3CCH3HOOH

CAS RN 2096336-24-0

IUPAC Name: {3-[(2,6-dimethylphenyl)carbamoyl]-5-fluorophenyl}boronic acid
Synonyms: 3-[(2,6-dimethylphenyl)carbamoyl]-5-fluorophenylboronic acid
Molecular Weight (g/mol): 287.1
Molecular Formula: C15H15BFNO3
InChi Key: OLDZXRPMUUYXQI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)C1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results