accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2096336-75-1

IUPAC Name: {2-fluoro-5-[(3-fluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[(3-fluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 277.03
Molecular Formula: C13H10BF2NO3
InChi Key: XUFYLZBKRBENSW-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results