accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHNFHOOH

CAS RN 2096337-03-8

IUPAC Name: {4-fluoro-3-[(2-fluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-fluoro-3-[(2-fluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 277.03
Molecular Formula: C13H10BF2NO3
InChi Key: UODKOFFESOCMSB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C(=O)NC2=CC=CC=C2F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results