CAS RN 2096337-08-3
IUPAC Name:
{3-[(4-bromo-3-methylphenyl)carbamoyl]-5-fluorophenyl}boronic acid
Synonyms:
3-[(4-bromo-3-methylphenyl)carbamoyl]-5-fluorophenylboronic acid
Molecular Weight (g/mol):
351.97
Molecular Formula:
C14H12BBrFNO3
InChi Key:
DIXOWPIMCKUEGL-UHFFFAOYSA-N
SMILES:
CC1=C(Br)C=CC(NC(=O)C2=CC(F)=CC(=C2)B(O)O)=C1
1
–
1
of
1
results
1
5-(4-Bromo-3-methylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%, Thermo Scientific™
CAS: 2096337-08-3 Molecular Formula: C14H12BBrFNO3 Molecular Weight (g/mol): 351.97 MDL Number: MFCD20040212 InChI Key: DIXOWPIMCKUEGL-UHFFFAOYSA-N Synonym: 5-4-bromo-3-methylphenylcarbamoyl-3-fluorobenzeneboronic acid PubChem CID: 73995851