accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH(E/Z)

CAS RN 20972-36-5

IUPAC Name: 4-(4-methylphenyl)-4-oxobut-2-enoic acid
Synonyms: 4-(4-methylphenyl)-4-oxobut-2-enoic acid
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: VNJMEFZKYZHWEO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C=CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results