accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2097998-59-7

IUPAC Name: 2-{2-[(20-{[1-(dicyanomethylidene)-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-2-ylidene]methyl}-12,12,24,24-tetrakis(4-hexylphenyl)-5,9,17,21-tetrathiaheptacyclo[13.9.0.0³,¹³.0⁴,¹¹.0⁶,¹⁰.0¹⁶,²³.0¹⁸,²²]tetracosa-1(15),2,4(11),6(10),7,13,16(23),18(22),19-nonaen-8-yl)methylidene]-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene}propanedinitrile
Synonyms: 2-{2-[(20-{[1-(dicyanomethylidene)-5,6-difluoro-3-oxoinden-2-ylidene]methyl}-12,12,24,24-tetrakis(4-hexylphenyl)-5,9,17,21-tetrathiaheptacyclo[13.9.0.0³,¹³.0⁴,¹¹.0⁶,¹⁰.0¹⁶,²³.0¹⁸,²²]tetracosa-1(15),2,4(11),6(10),7,13,16(23),18(22),19-nonaen-8-yl)methylidene]-5,6-difluoro-3-oxoinden-1-ylidene}propanedinitrile
Molecular Weight (g/mol): 1499.92
Molecular Formula: C94H78F4N4O2S4
InChi Key: JOZQXSUYCMNTCH-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C1(C2=CC3=C(C=C2C2=C1C1=C(S2)C=C(S1)C=C1C(=O)C2=CC(F)=C(F)C=C2C1=C(C#N)C#N)C(C1=C3SC2=C1SC(C=C1C(=O)C3=CC(F)=C(F)C=C3C1=C(C#N)C#N)=C2)(C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)