accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OHNHCl

CAS RN 209808-47-9

IUPAC Name: N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide hydrochloride
Synonyms: N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide hydrochloride
Molecular Weight (g/mol): 384.95
Molecular Formula: C23H29ClN2O
InChi Key: OTXTZCLQEGSAMW-UHFFFAOYSA-N
SMILES: Cl.CCN(CC)C(=O)C1=CC=C(C=C1)C(=C1CCNCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results