accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2H3C

CAS RN 209982-56-9

IUPAC Name: 2-ethyladamantan-2-yl 2-methylprop-2-enoate
Synonyms: 2-ethyladamantan-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 248.37
Molecular Formula: C16H24O2
InChi Key: DCTVCFJTKSQXED-UHFFFAOYSA-N
SMILES: CCC1(OC(=O)C(C)=C)C2CC3CC(C2)CC1C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)