Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-2-methacryloyloxyadamantane (stabilized with MEHQ) 97.0+%, TCI America™
SDP

Catalog No. E09095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E09095G Supplier TCI America Supplier No. E09095G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 209982-56-9
Molecular Formula C16H24O2
Molecular Weight (g/mol) 248.366
MDL Number MFCD06411144
InChI Key DCTVCFJTKSQXED-UHFFFAOYSA-N
Synonym 2-Ethyl-2-adamantyl Methacrylate, Methacrylic Acid 2-Ethyl-2-adamantyl Ester
PubChem CID 9813645
IUPAC Name (2-ethyl-2-adamantyl) 2-methylprop-2-enoate
SMILES CCC1(C2CC3CC(C2)CC1C3)OC(=O)C(=C)C

Specifications

Melting Point 40°C
Color White
Quantity 5 g
Formula Weight 248.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Ethyl-2-methacryloyloxyadamantane (stabilized with MEHQ)
EINECSNumber (4)-1880
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.