accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO

CAS RN 21005-45-8

IUPAC Name: 1-methyl-1H-indole-6-carbaldehyde
Synonyms: 1-methylindole-6-carbaldehyde
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: QGAKKGIWURKKHV-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=C(C=O)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results