accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COCH3CH3CH3H3CPOOOOPCH3H3CCH3OCH3CH3CH3H3CCH3CH3H3COH3CH3CCH3H3CH3CH3CCH3OH3CCH3H3CCH3(?)

CAS RN 210169-40-7

IUPAC Name: (4-{5-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane
Synonyms: (4-{5-[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphane
Molecular Weight (g/mol): 1179.55
Molecular Formula: C74H100O8P2
InChi Key: ZNORAFJUESSLTM-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1C(C)(C)C)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C1=CC(=C(OC)C(=C1)C(C)(C)C)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)