accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3CNOSiCH3CH3H3CSiCH3CH3H3C

CAS RN 21023-20-1

IUPAC Name: 2,2,5,5-tetramethyl-4-(trimethylsilyl)-3-oxa-4-aza-2,5-disilahexane
Synonyms: 2,2,5,5-tetramethyl-4-(trimethylsilyl)-3-oxa-4-aza-2,5-disilahexane
Molecular Weight (g/mol): 249.58
Molecular Formula: C9H27NOSi3
InChi Key: WEFDIXZMPUWSQQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)ON([Si](C)(C)C)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results