Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,O-Tris(trimethylsilyl)hydroxylamine 97.0+%, TCI America™
SDP

Catalog No. T16751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16751G Supplier TCI America Supplier No. T16751G

Chemical Identifiers

CAS 21023-20-1
Molecular Formula C9H27NOSi3
Molecular Weight (g/mol) 249.576
MDL Number MFCD01321190
InChI Key WEFDIXZMPUWSQQ-UHFFFAOYSA-N
PubChem CID 519835
IUPAC Name [dimethyl-[trimethylsilyl(trimethylsilyloxy)amino]silyl]methane
SMILES C[Si](C)(C)N(O[Si](C)(C)C)[Si](C)(C)C

Specifications

Melting Point -36°C
Color Yellow
Boiling Point 128°C
Quantity 1 g
UN Number 1760
Formula Weight 249.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,O-Tris(trimethylsilyl)hydroxylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.