CAS RN 210282-30-7
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(2-nitrophenyl)propanoic acid
Molecular Weight (g/mol):
432.43
Molecular Formula:
C24H20N2O6
InChi Key:
KVLRWXBYPKSBFY-NRFANRHFSA-N
SMILES:
OC(=O)[C@H](CC1=C(C=CC=C1)[N+]([O-])=O)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Sigma Aldrich N-(2-Bromoethyl)Phthalimide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.