accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNBr

CAS RN 2103-94-8

IUPAC Name: 4-(4-bromophenyl)-1,3-thiazol-2-amine
Synonyms: 4-(4-bromophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 255.13
Molecular Formula: C9H7BrN2S
InChi Key: ZBRNKOLWXWMLTA-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)