Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-(4-bromophenyl)thiazole 98.0+%, TCI America™
SDP

Catalog No. A28295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28295G Supplier TCI America Supplier No. A28295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2103-94-8
Molecular Formula C9H7BrN2S
Molecular Weight (g/mol) 255.133
MDL Number MFCD00051953
InChI Key ZBRNKOLWXWMLTA-UHFFFAOYSA-N
Synonym 2-amino-4-4-bromophenyl thiazole, 4-4-bromophenyl-1,3-thiazol-2-amine, 4-4-bromophenyl thiazol-2-amine, 4-4-bromophenyl-2-thiazolamine, 2-thiazolamine, 4-4-bromophenyl, 4-4-bromo-phenyl-thiazol-2-ylamine, 4-4-bromophenyl-1,3-thiazole-2-ylamine, 2-amino-4-p-bromophenyl thiazole, 2-amino-4-4-bromophenyl-1,3-thiazole, 2-amino-4-4'-bromophenyl-1,3-thiazole
PubChem CID 620683
IUPAC Name 4-(4-bromophenyl)-1,3-thiazol-2-amine
SMILES C1=CC(=CC=C1C2=CSC(=N2)N)Br

Specifications

Melting Point 186°C
Quantity 5 g
Formula Weight 255.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(4-bromophenyl)thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.