CAS RN 210353-53-0
IUPAC Name:
7-[(4Z)-3-(aminomethyl)-4-(methoxyimino)pyrrolidin-1-yl]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid; methanesulfonic acid
Synonyms:
gemifloxacin mesylate
Molecular Weight (g/mol):
485.49
Molecular Formula:
C19H24FN5O7S
InChi Key:
JIYMVSQRGZEYAX-CWUUNJJBNA-N
SMILES:
CS(O)(=O)=O.CO\N=C1/CN(CC1CN)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=N1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Gemifloxacin mesylate
Thermo Scientific Chemicals Gemifloxacin mesylate
CAS: 210353-53-0 Molecular Formula: C19H24FN5O7S