accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21085-72-3

IUPAC Name: methyl (2S,3S,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxane-2-carboxylate
Synonyms: methyl (2S,3S,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxane-2-carboxylate
Molecular Weight (g/mol): 397.17
Molecular Formula: C13H17BrO9
InChi Key: GWTNLHGTLIBHHZ-SVNGYHJRSA-N
SMILES: COC(=O)[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)