accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSBr

CAS RN 21101-63-3

IUPAC Name: 1-(bromomethyl)-4-[(trifluoromethyl)sulfanyl]benzene
Synonyms: 1-(bromomethyl)-4-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol): 271.1
Molecular Formula: C8H6BrF3S
InChi Key: VAYGJBCOCRVROJ-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=C(CBr)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)