Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethylthio)benzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. T23915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23915G 5 g
T23911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23915G Supplier TCI America Supplier No. T23915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21101-63-3
Molecular Formula C8H6BrF3S
Molecular Weight (g/mol) 271.095
MDL Number MFCD00129189
InChI Key VAYGJBCOCRVROJ-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide
PubChem CID 2777882
IUPAC Name 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1CBr)SC(F)(F)F

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 3261
Formula Weight 271.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethylthio)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.