accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 211115-05-8

IUPAC Name: chloro[2-(4-methoxyphenyl)ethyl]magnesium
Synonyms: chloro[2-(4-methoxyphenyl)ethyl]magnesium
Molecular Weight (g/mol): 194.94
Molecular Formula: C9H11ClMgO
InChi Key: BAZYZLFGOBVFJO-UHFFFAOYSA-M
SMILES: COC1=CC=C(CC[Mg]Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results