Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenethylmagnesium chloride, 0.5M solution in THF, AcroSeal™

Catalog No. AC440800500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440800500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440800500 Supplier Thermo Scientific Chemicals Supplier No. 440800500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 211115-05-8 | C9H11ClMgO | 194.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211115-05-8 , 109-99-9
Molecular Formula C9H11ClMgO
Molecular Weight (g/mol) 194.94
MDL Number MFCD01319915
InChI Key BAZYZLFGOBVFJO-UHFFFAOYSA-M
Synonym 4-methoxyphenethylmagnesium chloride, 4-methoxyphenethylmagnesium chloride solution, 2-4-methoxyphenyl ethylmagnesium chloride, 0.25m in 2-methf, 4-methoxyphenethylmagnesium chloride 0.5 m in tetrahydrofuran, 4-methoxyphenethylmagnesium chloride solution, 0.5 m in thf
PubChem CID 16217188
IUPAC Name magnesium;1-ethyl-4-methoxybenzene;chloride
SMILES COC1=CC=C(CC[Mg]Cl)C=C1

Specifications

Color Yellow to Brown
Density 0.9250g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula H3COC6H4(CH2)2MgCl
Quantity 50 mL
Specific Gravity 0.925
Solubility Information Solubility in water: reacts
Formula Weight 194.94
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenethylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.