accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21129-09-9

H3COHOH

CAS RN 21129-09-9

IUPAC Name: tetradecane-1,2-diol
Synonyms: myristyl glycol
Molecular Weight (g/mol): 230.39
Molecular Formula: C14H30O2
InChi Key: DWANEFRJKWXRSG-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC(O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results