Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Tetradecanediol 90.0+%, TCI America™
SDP

Catalog No. T135325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T135325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T135325G Supplier TCI America Supplier No. T135325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21129-09-9
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.392
MDL Number MFCD00009986
InChI Key DWANEFRJKWXRSG-UHFFFAOYSA-N
Synonym 1,2-Dihydroxytetradecane
PubChem CID 89436
ChEBI CHEBI:84951
IUPAC Name tetradecane-1,2-diol
SMILES CCCCCCCCCCCCC(CO)O

Specifications

Melting Point 67°C
Color White
Quantity 25 g
Formula Weight 230.39
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Tetradecanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.