accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2113-58-8

IUPAC Name: 3-nitro-1,1'-biphenyl
Synonyms: 3-nitrobiphenyl
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: FYRPEHRWMVMHQM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results