Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrodiphenyl 99.0+%, TCI America™
SDP

Catalog No. N019325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N019325G 25 g
N01931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N019325G Supplier TCI America Supplier No. N019325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2113-58-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD00007245
InChI Key FYRPEHRWMVMHQM-UHFFFAOYSA-N
Synonym 3-nitrobiphenyl, biphenyl, 3-nitro, m-nitrobiphenyl, 3-nitro-1,1'-biphenyl, 1,1'-biphenyl, 3-nitro, 3-nitrodiphenyl, 1,1'-biphenyl, 3-nitro-9ci, m-nitrodiphenyl, 3'-nitrobiphenyl, acmc-209fhv
PubChem CID 16450
IUPAC Name 1-nitro-3-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 61°C
Color Yellow
Boiling Point 204°C
Quantity 25 g
Formula Weight 199.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrodiphenyl
RTECSNumber DV5570000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.