CAS RN 211308-79-1
IUPAC Name:
3-iodo-5-methylpyridin-2-amine
Synonyms:
3-iodo-5-methylpyridin-2-amine
Molecular Weight (g/mol):
234.04
Molecular Formula:
C6H7IN2
InChi Key:
GSRKHYYLAZKHSA-UHFFFAOYSA-N
SMILES:
CC1=CC(I)=C(N)N=C1
1
–
1
of
1
results
1
Sigma Aldrich 3-Iodo-5-methyl-pyridin-2-ylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.