accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 2114-39-8

IUPAC Name: (2-bromopropyl)benzene
Synonyms: (2-bromopropyl)benzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: NVYOCAOZCSNIHR-UHFFFAOYNA-N
SMILES: CC(Br)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)