Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-phenylpropane 98%, Thermo Scientific™

Catalog No. AC107300050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC107300050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC107300050 Supplier Thermo Scientific Chemicals Supplier No. 107300050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2114-39-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000149
InChI Key NVYOCAOZCSNIHR-UHFFFAOYNA-N
Synonym 2-bromopropyl benzene, 2-bromo-1-phenylpropane, 1-phenyl-2-bromopropane, 2-bromo-propyl-benzene, benzene, 2-bromopropyl, 2-bromanylpropylbenzene, 2-bromopropyl-benzene, 1-2-bromopropyl benzene, acmc-1cr50
PubChem CID 102754
IUPAC Name (2-bromopropyl)benzene
SMILES CC(Br)CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Brown
Infrared Spectrum Authentic
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Linear Formula C6H5CH2CH(Br)CH3
Quantity 5 g
Beilstein 05, I, 190
Formula Weight 199.09
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Bromo-1-phenylpropane, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.