accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2

CAS RN 2114-42-3

IUPAC Name: (prop-2-en-1-yl)cyclohexane
Synonyms: allyl-cyclohexane
Molecular Weight (g/mol): 124.23
Molecular Formula: C9H16
InChi Key: KVOZXXSUSRZIKD-UHFFFAOYSA-N
SMILES: C=CCC1CCCCC1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)