Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylcyclohexane 97.0+%, TCI America™
SDP

Catalog No. A120125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A120125ML 25 mL
A12015ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A120125ML Supplier TCI America Supplier No. A120125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2114-42-3
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00013769
InChI Key KVOZXXSUSRZIKD-UHFFFAOYSA-N
Synonym allylcyclohexane, allyl-cyclohexane, cyclohexane, allyl, 3-cyclohexyl-1-propene, cyclohexane, 2-propenyl, 1-cyclohexyl-2-propene, 1-propene, 3-cyclohexyl, allyl cyclohexane, 3-cyclohexylpropene, 2-propenyl-cyclohexane
PubChem CID 75027
IUPAC Name prop-2-enylcyclohexane
SMILES C=CCC1CCCCC1

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.