accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiOOCH3CH2OCH3OCH3

CAS RN 21142-29-0

IUPAC Name: 3-(triethoxysilyl)propyl 2-methylprop-2-enoate
Synonyms: 3-(triethoxysilyl)propyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 290.43
Molecular Formula: C13H26O5Si
InChi Key: URDOJQUSEUXVRP-UHFFFAOYSA-N
SMILES: CCO[Si](CCCOC(=O)C(C)=C)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results