Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Triethoxysilyl)propyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™
SDP

Catalog No. T267625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T267625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. T267625G Supplier TCI America Supplier No. T267625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 21142-29-0
Molecular Formula C13H26O5Si
Molecular Weight (g/mol) 290.431
MDL Number MFCD00069153
InChI Key URDOJQUSEUXVRP-UHFFFAOYSA-N
Synonym Triethoxy(3-methacryloyloxypropyl)silane, Methacrylic Acid 3-(Triethoxysilyl)propyl Ester
PubChem CID 88798
IUPAC Name 3-triethoxysilylpropyl 2-methylprop-2-enoate
SMILES CCO[Si](CCCOC(=O)C(=C)C)(OCC)OCC

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
Formula Weight 290.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Triethoxysilyl)propyl Methacrylate (stabilized with BHT)
EINECSNumber (2)-2076
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.