accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3CNOFFFSiCH3CH3H3C

CAS RN 21149-38-2

IUPAC Name: 2,2,2-trifluoro-N,N-bis(trimethylsilyl)acetamide
Synonyms: 2,2,2-trifluoro-N,N-bis(trimethylsilyl)acetamide
Molecular Weight (g/mol): 257.4
Molecular Formula: C8H18F3NOSi2
InChi Key: RZYHXKLKJRGJGP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N(C(=O)C(F)(F)F)[Si](C)(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results