Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-O-bis-(Trimethylsilyl)trifluoroacetamide, 99.26%, MP Biomedicals™

Catalog No. ICN15047605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15047605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15047605 Supplier MP Biomedicals, Inc Supplier No. 0215047605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-O-bis-(Trimethylsilyl)trifluoroacetamide is a silylating agent for amino acids, carbohydrates, and steroids.

  • N-O-bis-(Trimethylsilyl)trifluoroacetamide is a derivatization reagent employed in a GC-MS analysis of furfural content in foods.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21149-38-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
MDL Number MFCD00008269
InChI Key RZYHXKLKJRGJGP-UHFFFAOYSA-N
Synonym bis trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n,n-bis trimethylsilyl acetamide, acetamide, 2,2,2-trifluoro-n,n-bis trimethylsilyl, unii-5t8na426kz, n,n-bstfa, ksc497c2d, bistrimethylsilyltrifluoroacetamide, bis-trimethylsilyltrifluoroacetamide, bistrimethylsilyl-trifluoroacetamide
PubChem CID 65279
IUPAC Name 2,2,2-trifluoro-N,N-bis(trimethylsilyl)acetamide
SMILES C[Si](C)(C)N(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 257.4
Grade Not Available
Physical Form Liquid
Chemical Name or Material N-O-bis-(Trimethylsilyl)trifluoroacetamide, FTIR
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.