accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOCl

CAS RN 21151-56-4

IUPAC Name: 1-chloro-4-(chloromethoxy)benzene
Synonyms: 1-chloro-4-(chloromethoxy)benzene
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: PJLGIZXAYTZSIZ-UHFFFAOYSA-N
SMILES: ClCOC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results