Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,4-Dichloroanisole, 97%, Thermo Scientific™

Catalog No. AC170160250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC170160250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170160250 Supplier Thermo Scientific Chemicals Supplier No. 170160250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21151-56-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H6Cl2O
Linear Formula ClC6H4OCH2Cl
MDL Number MFCD00000883
Synonym alpha,4-dichloroanisole, 1-chloro-4-chloromethoxy benzene, .alpha.,para-dichloroanisole, benzene, 1-chloro-4-chloromethoxy, a,4-dichloroanisole, .alpha.,4-dichloroanisole, 4-chlorophenoxychloromethane, alpha-chloro-p-chloroanisole, p-chlorophenoxymethyl chloride, 4-chlorophenoxymethyl chloride
InChI Key PJLGIZXAYTZSIZ-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OCCl)Cl
IUPAC Name 1-chloro-4-(chloromethoxy)benzene
Molecular Weight (g/mol) 177.024
PubChem CID 88801
Formula Weight 177.03
Percent Purity ≥96.0%
Chemical Name or Material α, 4-Dichloroanisole, 97%
Show More Show Less
EINECSNumber 244-246-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.