accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHONOO

CAS RN 21161-11-5

IUPAC Name: 2-nitrobenzene-1,3-dicarboxylic acid
Synonyms: 2-nitrobenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 211.13
Molecular Formula: C8H5NO6
InChi Key: CAHWDGJDQYAFHM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(C(O)=O)=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results