Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitroisophthalic Acid 95.0+%, TCI America™
SDP

Catalog No. N10865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10865G Supplier TCI America Supplier No. N10865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21161-11-5
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00017007
InChI Key CAHWDGJDQYAFHM-UHFFFAOYSA-N
PubChem CID 88806
SMILES C1=CC(=C(C(=C1)C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 295°C
Color White-Yellow
Quantity 5 g
Formula Weight 211.13
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitroisophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.