CAS RN 21165-77-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl N-4-[2-(5-chloro-2-methoxybenzamido)ethyl]benzenesulfonylcarbamate, MilliporeSigma™ Supelco™
This British Pharmacopoeia (BP) Reference Standard is provided as delivered and specified by the issuing Pharmacopoeia. All supporting information, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.