CAS RN 212-74-8
IUPAC Name:
tetraphenylene
Synonyms:
tetraphenylene
Molecular Weight (g/mol):
304.39
Molecular Formula:
C24H16
InChi Key:
KTQYWNARBMKMCX-UHFFFAOYSA-N
SMILES:
C1=CC2=C3C=CC=CC3=C3C=CC=CC3=C3C=CC=CC3=C2C=C1
1
–
1
of
1
results
1
Tetraphenylene 98%, Thermo Scientific™
CAS: 212-74-8 Molecular Formula: C24H16 Molecular Weight (g/mol): 304.392 InChI Key: KTQYWNARBMKMCX-UHFFFAOYSA-N Synonym: 4az,8az,12az,16az-tetraphenylene PubChem CID: 2724868 IUPAC Name: tetraphenylene SMILES: C1=CC2=C3C=CC=CC3=C4C=CC=CC4=C5C=CC=CC5=C2C=C1