accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 21210-43-5

IUPAC Name: methyl 2-hydroxy-2-phenylacetate
Synonyms: methyl 2-hydroxy-2-phenylacetate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: ITATYELQCJRCCK-UHFFFAOYNA-N
SMILES: COC(=O)C(O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)