Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(+)-mandelate, 99%, Thermo Scientific™

Catalog No. AAL1407003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1407003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1407003 Supplier Thermo Scientific Chemicals Supplier No. L1407003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21210-43-5
Melting Point 56°C to 58°C
Flash Point >110°C (230°F)
Molecular Formula C9H10O3
Quantity 1 g
MDL Number MFCD00064246
Beilstein 2614741
Synonym methyl s-+-mandelate, s-+-methyl mandelate, s-methyl 2-hydroxy-2-phenylacetate, methyl 2s-2-hydroxy-2-phenylacetate, s-methyl mandelate, l-+-mandelic acid methyl ester, +-methyl l-mandelate, methyl l-+-mandelate, methyl hydroxy phenyl acetate #, methyl s-mandelate
InChI Key ITATYELQCJRCCK-UHFFFAOYNA-N
SMILES COC(=O)C(O)C1=CC=CC=C1
Optical Rotation +144° (c= 1 in Methanol)
IUPAC Name methyl (2S)-2-hydroxy-2-phenylacetate
Molecular Weight (g/mol) 166.18
PubChem CID 643570
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl (S)-(+)-mandelate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.