accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3INHOHNOFFF

CAS RN 212631-61-3

IUPAC Name: N-(cyclopropylmethoxy)-3,4,5-trifluoro-2-[(4-iodo-2-methylphenyl)amino]benzamide
Synonyms: N-(cyclopropylmethoxy)-3,4,5-trifluoro-2-[(4-iodo-2-methylphenyl)amino]benzamide
Molecular Weight (g/mol): 476.24
Molecular Formula: C18H16F3IN2O2
InChi Key: UHAXDAKQGVISBZ-UHFFFAOYSA-N
SMILES: CC1=C(NC2=C(F)C(F)=C(F)C=C2C(=O)NOCC2CC2)C=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results