accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212755-84-5

IUPAC Name: 2-aminocyclopentane-1-carboxylic acid hydrochloride
Synonyms: 2-aminocyclopentane-1-carboxylic acid hydrochloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: LVBDVNLIEHCCTP-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)