Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99%, Thermo Scientific™

Catalog No. AC298382500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC298382500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC298382500 Supplier Thermo Scientific Chemicals Supplier No. 298382500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 212755-84-5
Molecular Formula HCl·1/2H2O
Molecular Weight (g/mol) 174.63
MDL Number MFCD00800502
InChI Key LVBDVNLIEHCCTP-JBUOLDKXSA-N
Synonym 1s,2r-2-aminocyclopentane-1-carboxylic acid hydrate hydrochloride, cis-2-amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99
PubChem CID 2733976
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxylic acid;hydrochloride
SMILES C1CC(C(C1)N)C(=O)O.Cl

Specifications

Melting Point 199°C to 202°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water
Quantity 250 mg
Formula Weight 174.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.