accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21282-97-3

IUPAC Name: 2-[(2-methylprop-2-enoyl)oxy]ethyl 3-oxobutanoate
Synonyms: hema acetoacetate
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: IBDVWXAVKPRHCU-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)OCCOC(=O)C(C)=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)