Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Methacryloyloxy)ethyl acetoacetate

Catalog No. 5374034L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-740-34L 4 L
53-740-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-740-34L Supplier Sigma Aldrich Supplier No. 5374034L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(methacryloyloxy)ethyl acetoacetate is a methylene active compound that may be used in the synthesis of 2-(methacryloyloxy)ethyl 6-methyl-2-oxo-4-phenyl-1,2,3,4- tetrahydropyrimidine-5-carboxylate
  • It may be used as a polymerizable chelating agent for metal alkoxides to form polymerizable oxide precursors

Specifications

CAS 21282-97-3
Density 1.122 g/mL (at 25°C (literature))
Boiling Point 100°C (0.8 mmHg)
Molecular Formula C10H14O5
Refractive Index n20/D 1.456 (literature)
Linear Formula CH3COCH2CO2CH2CH2O2CC(CH3)=CH2
MDL Number MFCD00054405
Quantity 4 L
Synonym AAEM; (2-Acetoacetoxy)ethyl methacrylate; Ethylene glycol monoacetoacetate monomethacrylate
Molecular Weight (g/mol) 214.22
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.