accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 213-46-7

IUPAC Name: picene
Synonyms: picene dibenzo(a,i)phenanthrene 1,2:7,8-dibenzophenanthrene 3,4-benzchrysene benzo(a)chrysene β,β-binaphthyleneethene 1,2,7,8-dibenzphenanthrene pycene
Molecular Weight (g/mol): 278.35
Molecular Formula: C22H14
InChi Key: GBROPGWFBFCKAG-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=C4C=CC5=CC=CC=C5C4=CC=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results